Merck
  • Molecularly imprinted polymer based microtiter chemiluminescence array for determination of phenothiazines and benzodiazepines in pork.
Molecularly imprinted polymer based microtiter chemiluminescence array for determination of phenothiazines and benzodiazepines in pork.
Analytical biochemistry (2018-05-29)
Wan Qiu Xia, Jun Huang, Geng Nan Wang, Jing Liu, Jian Ping Wang
ABSTRACT
In this study, a molecularly imprinted polymer based chemiluminescence array capable of simultaneous determining phenothiazines and benzodiazepines was first reported. Two polymers were coated in different wells of the conventional 96-well microtiter plate as the recognition reagents, and the added analytes competed with a horseradish peroxidase-labeled bi-hapten conjugate to bind the recognition reagents. The light signal was induced by using a highly effective luminol-H2O2-IMP system. The assay procedure consisted of only one sample-loading step prior to data acquisition. Then, the array was used to determine 4 phenothiazines and 5 benzodiazepines in pork simultaneously. The limits of detection for the 9 drugs were in a range of 0.001-0.01 ng/mL, and the recoveries from the fortified blank pork were in a range of 63.5%-94.1%. Furthermore, the array could be reused for 8 times. The detection results for some real pork samples were consistent with an ultra performance liquid chromatography method.
Sigma-Aldrich
4-(Imidazol-1-yl)phenol, 97%