- ETS Exposure and PAH Body Burden in Nonsmoking Italian Adults.
ETS Exposure and PAH Body Burden in Nonsmoking Italian Adults.
International journal of environmental research and public health (2018-06-06)
Laura Campo, Elisa Polledri, Petra Bechtold, Giulia Gatti, Giulia Quattrini, Luca Olgiati, Michael Romolo, Andrea Ranzi, Paolo Lauriola, Giuliano Carrozzi, Silvia Fustinoni
PMID29865209
ABSTRACT
Active smoking is associated with increased body burden of polycyclic aromatic hydrocarbons (PAHs); the aim of this study was to assess whether environmental tobacco smoking (ETS) increases the internal dose of PAHs. In 344 nonsmoking Italian adults, out of 497 individuals selected as representative of the population of the town of Modena, ETS exposure was evaluated by a self-administered questionnaire and by the measurement of urinary cotinine (COT-U). PAH exposure was assessed by the measurement of urinary 1-hydroxypyrene (1-OHPYR) and of ten urinary PAHs. In all subjects, median (5th⁻95th percentile) COT-U was 0.47 (.