Merck
  • ICE-LAP6, a novel member of the ICE/Ced-3 gene family, is activated by the cytotoxic T cell protease granzyme B.
ICE-LAP6, a novel member of the ICE/Ced-3 gene family, is activated by the cytotoxic T cell protease granzyme B.
The Journal of biological chemistry (1996-07-12)
H Duan, K Orth, A M Chinnaiyan, G G Poirier, C J Froelich, W W He, V M Dixit
ABSTRACT
Members of the ICE/Ced-3 gene family are likely effector components of the cell death machinery. Here, we characterize a novel member of this family designated ICE-LAP6. By phylogenetic analysis, ICE-LAP6 is classified into the Ced-3 subfamily which includes Ced-3, Yama/CPP32/apopain, Mch2, and ICE-LAP3/Mch3/CMH-1. Interestingly, ICE-LAP6 contains an active site QACGG pentapeptide, rather than the QACRG pentapeptide shared by other family members. Overexpression of ICE-LAP6 induces apoptosis in MCF7 breast carcinoma cells. More importantly, ICE-LAP6 is proteolytically processed into an active cysteine protease by granzyme B, an important component of cytotoxic T cell-mediated apoptosis. Once activated, ICE-LAP6 is able to cleave the death substrate poly(ADP-ribose) polymerase into signature apoptotic fragments.
Sigma-Aldrich
Caspase 9 human, ≥90% (SDS-PAGE), recombinant, expressed in E. coli (C-terminal histidine-tagged), buffered aqueous solution, >2,000 units/mg protein