Merck
  • A new virus, classifiable in the family Tombusviridae, found infecting Solanum tuberosum in the UK.
A new virus, classifiable in the family Tombusviridae, found infecting Solanum tuberosum in the UK.
Archives of virology (2018-03-02)
Wendy Monger, Colin Jeffries
ABSTRACT
A novel virus was discovered in a freeze-dried collection held at SASA, UK, originating from potato (Solanum tuberosum) cv. Nadine. The complete sequence of the viral RNA was determined to be 3674 nucleotides in length encoding five predicted proteins. Based on the deduced genome organization and phylogenetic analysis, this virus represents a putative new member of the genus Alphanecrovirus, family Tombusviridae, most closely related to isolates of Olive mild mosaic virus. The virus was easily transmitted to indicator plants with symptoms that were slower to develop and less severe than those of related viruses. To distinguish this virus, the clearest symptom differences occurred with Nicotiana debneyi, Chenopodium amaranticolor and Ch. quinoa. The virus was detected with antisera to the related viruses tobacco necrosis virus A and tobacco necrosis virus D. The close association to the tobacco necrosis viruses would suggest this virus is not a new introduction to potato but in the past has been misidentified as one of these viruses. The virus isolate has been named potato necrosis virus.