Merck
  • Intravital imaging of a pulmonary endothelial surface layer in a murine sepsis model.
Intravital imaging of a pulmonary endothelial surface layer in a murine sepsis model.
Biomedical optics express (2018-05-16)
Inwon Park, Kibaek Choe, Howon Seo, Yoonha Hwang, Eunjoo Song, Jinhyo Ahn, You Hwan Jo, Pilhan Kim
ABSTRACT
Direct intravital imaging of an endothelial surface layer (ESL) in pulmonary microcirculation could be a valuable approach to investigate the role of a vascular endothelial barrier in various pathological conditions. Despite its importance as a marker of endothelial cell damage and impairment of the vascular system, in vivo visualization of ESL has remained a challenging technical issue. In this work, we implemented a pulmonary microcirculation imaging system integrated to a custom-design video-rate laser scanning confocal microscopy platform. Using the system, a real-time cellular-level microscopic imaging of the lung was successfully performed, which facilitated a clear identification of individual flowing erythrocytes in pulmonary capillaries. Subcellular level pulmonary ESL was identified in vivo by fluorescence angiography using a dextran conjugated fluorophore to label blood plasma and the red blood cell (RBC) exclusion imaging analysis. Degradation of ESL width was directly evaluated in a murine sepsis model in vivo, suggesting an impairment of pulmonary vascular endothelium and endothelial barrier dysfunction.