Merck
  • SNAI1, an endothelial-mesenchymal transition transcription factor, promotes the early phase of ocular neovascularization.
SNAI1, an endothelial-mesenchymal transition transcription factor, promotes the early phase of ocular neovascularization.
Angiogenesis (2018-04-21)
Jia-Xing Sun, Tian-Fang Chang, Man-Hong Li, Li-Juan Sun, Xian-Chun Yan, Zi-Yan Yang, Yuan Liu, Wen-Qin Xu, Yang Lv, Jing-Bo Su, Liang Liang, Hua Han, Guo-Rui Dou, Yu-Sheng Wang
ABSTRACT
Ocular neovascularization is a comprehensive process involved in retinal vascular development and several blinding diseases such as age-related macular degeneration and retinopathy of prematurity, with vascular endothelial growth factor (VEGF) regarded as the master regulator. However, the qualified effect of anti-VEGF therapy reveals that the underlying mechanisms are still not clearly identified. To initialize angiogenesis, endothelial cells undergo a phenotype switching to generate highly migratory and invasive cells. This process shares certain similar characters observed in endothelial-mesenchymal transition (EndMT). Here, we found that SNAI1, an EndMT transcription factor, was expressed by endothelial cells in both physiological and pathological ocular neovascularization. SNAI1 overexpression triggered cell morphological change and enhanced cell motility, while loss of SNAI1 attenuated migration, invasion and sprouting. RNA sequence analysis further revealed that SNAI1 knockdown decreased the expression of genes related to cytoskeleton rearrangement and ECM remodeling. Moreover, intravitreal injection of small interfering RNA of SNAI1 suppressed new vessel formation in developing retina as well as mice model of choroidal neovascularization and oxygen-induced retinopathy. Therefore, we propose that the EndMT transcription factor SNAI1 promotes the early phase of ocular neovascularization and may provide a potential therapeutic target.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-SNAI1 Antibody, clone 10H4.1, clone 10H4.1, from mouse