Skip to Content
Merck
TSKgel® SCX (H+) HPLC Column phase sulfonic acid, L × I.D. 30 cm × 7.8 mm, 5 μm particle size

TSKgel® SCX (H+) (5 µm) HPLC Column

TSKgel® SCX (H+) HPLC Column phase sulfonic acid, L × I.D. 30 cm × 7.8 mm, 5 μm particle size
Synonym(s):
TSKgel® SCX HPLC Column

Products

Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Technique
Showing 1-1 of 1