Skip to Content
Merck
Thymol Blue sodium salt ACS reagent, Dye content 95 %

Thymol Blue sodium salt

Thymol Blue sodium salt ACS reagent, Dye content 95 %
Synonym(s):
Thymolsulfonphthalein sodium salt
Empirical Formula (Hill Notation):
C27H29NaO5S
CAS Number:
Molecular Weight:
488.57
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Chemical Composition
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing