Tembotrione metabolite AE 1417268
Synonym(s):
2-{2-Chloro-4-mesyl-3-[(2,2,2-trifluoroethoxy)methyl]benzoyl}-4,6-dihydroxycyclohexane-1,3-dione
Empirical Formula (Hill Notation):
C17H16ClF3O8S
CAS Number:
Molecular Weight:
472.82
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing