Skip to Content
RU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Screw caps without O-rings
Products
Product Category
caps (12)
Color
Manufacturer
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
AXYSCA
Screw caps without O-rings,
assorted colors, pkg of 8x500caps/cs
Expand
View Pricing
AXYSCB
Screw caps without O-rings,
blue, pkg of 8x500caps/cs
Expand
View Pricing
AXYSCBR
Screw caps without O-rings,
brown, pkg of 8x500caps/cs
Expand
View Pricing
AXYSCC
Screw caps without O-rings,
clear, pkg of 8x500caps/cs
Expand
View Pricing
AXYSCG
Screw caps without O-rings,
green, pkg of 8x500caps/cs
Expand
View Pricing
AXYSCGR
Screw caps without O-rings,
gray, pkg of 8x500caps/cs
Expand
View Pricing
AXYSCO
Screw caps without O-rings,
orange, pkg of 8x500caps/cs
Expand
View Pricing
AXYSCP
Screw caps without O-rings,
pink, pkg of 8x500caps/cs
Expand
View Pricing
AXYSCR
Screw caps without O-rings,
red, pkg of 8x500caps/cs
Expand
View Pricing
AXYSCV
Screw caps without O-rings,
violet, pkg of 8x500caps/cs
Expand
View Pricing
AXYSCW
Screw caps without O-rings,
white, pkg of 8x500caps/cs
Expand
View Pricing
AXYSCY
Screw caps without O-rings,
yellow, pkg of 8x500caps/cs
Expand
View Pricing