Skip to Content
Merck
Salicylamide puriss., ≥99.0% (T)

Salicylamide

Salicylamide puriss., ≥99.0% (T)
Synonym(s):
2-Hydroxybenzamide, Salicylamide
Linear Formula:
2-(HO)C6H4CONH2
CAS Number:
Molecular Weight:
137.14
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing