Skip to Content
Merck
Oxybutynin chloride meets EP, USP testing specifications

Oxybutynin chloride

Oxybutynin chloride meets EP, USP testing specifications
Synonym(s):
α-Phenylcyclohexaneglycolic acid 4-(diethylamino)-2-butynyl ester hydrochloride, Oxybutynin chloride
Empirical Formula (Hill Notation):
C22H31NO3 · HCl
CAS Number:
Molecular Weight:
393.95
EC Number:

Products

Product Category
Brand
Color
Feature
Formula Weight
Gene Alias
Physical Form
Purity
Agency
Research Area
Quality Segment
Application
Package Size
API Family
Special Grade