
Omeprazole Impurity H

Synonym(s):
2-[(RS)-[(4-Chloro-3,5-dimethylpyridin-2-yl)methyl]sulfinyl]-5-methoxy-1H-benzimidazole, 2-[[(4-Chloro-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl]-6-methoxy-1H-benzimidazole, 4-Desmethoxy-4-chloro Omeprazole
Empirical Formula (Hill Notation):
C16H16ClN3O2S
CAS Number:
Molecular Weight:
349.84
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing