Skip to Content
Merck
Monoclonal Anti-Phosphotyrosine−Agarose antibody produced in mouse clone PT-66, purified immunoglobulin, PBS solution

Monoclonal Anti-Phosphotyrosine−Agarose antibody produced in mouse

Monoclonal Anti-Phosphotyrosine−Agarose antibody produced in mouse clone PT-66, purified immunoglobulin, PBS solution
Synonym(s):
Monoclonal Anti-Phosphotyrosine

Products

Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Recombinant Host
Antibody Form
Conjugate
Post Translational Modification
Technique