![Monoclonal Anti-Leukemia Inhibitory Factor antibody produced in mouse clone 9824, purified immunoglobulin, lyophilized powder](/deepweb/assets/sigmaaldrich/product/images/423/271/27902592-041b-49fc-9985-daa0be1192c4/640/27902592-041b-49fc-9985-daa0be1192c4.jpg)
Monoclonal Anti-Leukemia Inhibitory Factor antibody produced in mouse
![Monoclonal Anti-Leukemia Inhibitory Factor antibody produced in mouse clone 9824, purified immunoglobulin, lyophilized powder](/deepweb/assets/sigmaaldrich/product/images/423/271/27902592-041b-49fc-9985-daa0be1192c4/640/27902592-041b-49fc-9985-daa0be1192c4.jpg)
Synonym(s):
Anti-CDF, Anti-Cholinergic differentiation factor, Anti-HILDA, Anti-LIF
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing