Skip to Content
Merck
vibrant-m

Methylprednisolone acetate for system suitability

vibrant-m
Synonym(s):
11β,17,21-Trihydroxy-6α-methylpregna-1,4-diene-3,20-dione 21-acetate
Empirical Formula (Hill Notation):
C24H32O6
CAS Number:
Molecular Weight:
416.51

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1