Skip to Content
Merck
L-Lysine-13C6,15N2 hydrochloride 99 atom % 13C, 99 atom % 15N, 95% (CP)

L-Lysine-13C6,15N2 hydrochloride

L-Lysine-13C6,15N2 hydrochloride 99 atom % 13C, 99 atom % 15N, 95% (CP)
Synonym(s):
(S)-2,6-Diaminohexanoic acid-13C6,15N2 hydrochloride, 13C and 15N Labeled Lys, 13C and 15N Labeled lysine hydrochloride
Linear Formula:
H215N(13CH2)413CH(15NH2)13CO2H · HCl
CAS Number:
Molecular Weight:
190.59

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing