Kasugamycin hydrochloride
Synonym(s):
3-O-[2-Amino-4-[(carboxyiminomethyl)amino]-2,3,4,6-tetradeoxy-D-arabino-hexopyranosyl]-D-chiro-inositol hydrochloride
Empirical Formula (Hill Notation):
C14H25N3O9 · HCl
CAS Number:
Molecular Weight:
415.82
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing