![Isobavachalcone phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/203/486/9f2fa6b5-ad6b-497c-8b6e-a83ebbe2ce5f/640/9f2fa6b5-ad6b-497c-8b6e-a83ebbe2ce5f.png)
Isobavachalcone
![Isobavachalcone phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/203/486/9f2fa6b5-ad6b-497c-8b6e-a83ebbe2ce5f/640/9f2fa6b5-ad6b-497c-8b6e-a83ebbe2ce5f.png)
Synonym(s):
(2E)-1-[2,4-Dihydroxy-3-(3-methyl-2-butenyl)phenyl]-3-(4-hydroxyphenyl)-2-propen-1-one, 2′,4,4′-Trihydroxy-3′-(3-methyl-2-butenyl)-chalcone, Corylifolinin, Isobacachalcone
Empirical Formula (Hill Notation):
C20H20O4
CAS Number:
Molecular Weight:
324.37
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing