Skip to Content
Merck
Irbesartan British Pharmacopoeia (BP) Reference Standard

Irbesartan

Irbesartan British Pharmacopoeia (BP) Reference Standard
Synonym(s):
2-Butyl-3-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-1,3-diazaspiro[4.4]non-1-en-4-one
Empirical Formula (Hill Notation):
C25H28N6O
CAS Number:
Molecular Weight:
428.53

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique