Skip to Content
Merck
Histamine European Pharmacopoeia (EP) Reference Standard

Histamine

Histamine European Pharmacopoeia (EP) Reference Standard
Synonym(s):
2-(4-Imidazolyl)ethylamine
Empirical Formula (Hill Notation):
C5H9N3
CAS Number:
Molecular Weight:
111.15

Products

Product Category
Brand
Boiling Point (°C)
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Product Line
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique