Skip to Content
Merck
Haloxyfop-P certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Haloxyfop-P

Haloxyfop-P certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
(R)-2-[4-(3-Chloro-5-(trifluoromethyl)-2-pyridyloxy)phenoxy]propionic acid
Empirical Formula (Hill Notation):
C15H11ClF3NO4
CAS Number:
Molecular Weight:
361.70

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Reference Material Type
Special Grade