![Glucoerucin potassium salt phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/302/069/22b335bd-49c1-41f6-9ae5-0c7891319ba6/640/22b335bd-49c1-41f6-9ae5-0c7891319ba6.png)
Glucoerucin potassium salt
![Glucoerucin potassium salt phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/302/069/22b335bd-49c1-41f6-9ae5-0c7891319ba6/640/22b335bd-49c1-41f6-9ae5-0c7891319ba6.png)
Synonym(s):
4-Methylthiobutylglucosinolate potassium salt
Empirical Formula (Hill Notation):
C12H22KNO9S3
CAS Number:
Molecular Weight:
459.60
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing