![Ethyl acetate/Ethanol 3:1 (v/v) solution (Ethyl acetate solution with 26.2% v/v SDA 35A), suitable for HPLC](/deepweb/assets/sigmaaldrich/product/images/122/577/6c363cb7-29a9-4f9a-aabd-51c79f4093b5/640/6c363cb7-29a9-4f9a-aabd-51c79f4093b5.jpg)
Ethyl acetate/Ethanol 3:1 (v/v) solution
![Ethyl acetate/Ethanol 3:1 (v/v) solution (Ethyl acetate solution with 26.2% v/v SDA 35A), suitable for HPLC](/deepweb/assets/sigmaaldrich/product/images/122/577/6c363cb7-29a9-4f9a-aabd-51c79f4093b5/640/6c363cb7-29a9-4f9a-aabd-51c79f4093b5.jpg)
Synonym(s):
Ethyl acetate solution, Ethyl acetate solution, 3:1 (v:v) ethyl acetate: 200 proof ethanol
Linear Formula:
3 EtOAc/EtOH
Products
Product Category
Brand
Sustainability
Feature
Physical Form
Format
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing