Diclofenac Impurity D
Synonym(s):
2-{2-[(2-Bromo-6-chlorophenyl)amino]phenyl}acetic acid, 2-[(2-Bromo-6-chlorophenyl)amino]benzeneacetic acid
Empirical Formula (Hill Notation):
C14H11BrClNO2
CAS Number:
Molecular Weight:
340.60
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing