Skip to Content
RU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cytosine β-D-arabinofuranoside
Synonym(s):
(β-D-Arabinofuranosyl)cytosine, Ara-C, Arabinocytidine, Arabinosylcytosine, Cytarabine, Cytosine arabinoside
Empirical Formula (Hill Notation):
C
9
H
13
N
3
O
5
CAS Number:
147-94-4
Molecular Weight:
243.22
EC Number:
205-705-9
Products
Product Category
antineoplastics (1)
bioactive small molecules (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Gene Alias
Product Line
Research Area
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
V900339
Cytosine β-D-arabinofuranoside,
Vetec
™
, reagent grade, 90%
Expand
View Pricing