
Cyanidin 3-sophoroside chloride

Synonym(s):
Cyanidin 3-O-(2-O-β-D-glucopyranosyl)-β-D-glucopyranoside chloride, Cyanidin 3-diglucoside, Sophoroside
Empirical Formula (Hill Notation):
C27H31ClO16
CAS Number:
Molecular Weight:
646.98
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing