
Cladribine

Synonym(s):
2-Chloro-2′-deoxyadenosine, CdA, Cladribine
Empirical Formula (Hill Notation):
C10H12ClN5O3
CAS Number:
Molecular Weight:
285.69
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing