
Chiralyst Ru637

Synonym(s):
RuCl(p-cymene)[(S,S)-Ts-DPEN], [N-[(1S,2S)-2-(Amino-κN)-1,2-diphenylethyl]-4-methylbenzenesulfonamidato-κN]chloro[(1,2,3,4,5,6-η)-1-methyl-4-(1-methylethyl)benzene]-ruthenium
Empirical Formula (Hill Notation):
C31H35ClN2O2RuS
CAS Number:
Molecular Weight:
636.21
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing