Skip to Content
Merck
vibrant-m

Calcium Folinate Hydrate crystalline / Leucovorin Calcium crystalline

vibrant-m
Synonym(s):
Folinic acid calcium salt, Calcium folinate
Empirical Formula (Hill Notation):
C20H21CaN7O7
CAS Number:
Molecular Weight:
511.50

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Agency
Quality Segment
Application
Showing 1-1 of 1