![Aloin B phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/159/858/0274c9cd-c634-4483-8807-e09b4c86f9a2/640/0274c9cd-c634-4483-8807-e09b4c86f9a2.png)
Aloin B
![Aloin B phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/159/858/0274c9cd-c634-4483-8807-e09b4c86f9a2/640/0274c9cd-c634-4483-8807-e09b4c86f9a2.png)
Synonym(s):
Isobarbaloin
Empirical Formula (Hill Notation):
C21H22O9
CAS Number:
Molecular Weight:
418.39
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing