Skip to Content
Merck
α-Ionone analytical standard

α-Ionone

α-Ionone analytical standard
Synonym(s):
4-(2,6,6-Trimethyl-2-cyclohexenyl)-3-buten-2-one
Empirical Formula (Hill Notation):
C13H20O
CAS Number:
Molecular Weight:
192.30
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing