Skip to Content
Merck
3-Heptanone analytical standard

3-Heptanone

3-Heptanone analytical standard
Synonym(s):
Butyl ethyl ketone, Ethyl butyl ketone
Linear Formula:
CH3(CH2)3COC2H5
CAS Number:
Molecular Weight:
114.19
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing