
2-Methylglutaric-4,5-13C2 acid solution

Empirical Formula (Hill Notation):
13C2C4H10O4
Molecular Weight:
148.13
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Isotope
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing