![2′-Deoxyuridine 5′-triphosphate sodium salt solution 100 mM in H2O](/deepweb/assets/sigmaaldrich/product/structures/352/977/678c0cf4-b036-4984-b198-5f2a4dda679c/640/678c0cf4-b036-4984-b198-5f2a4dda679c.png)
2′-Deoxyuridine 5′-triphosphate sodium salt solution
![2′-Deoxyuridine 5′-triphosphate sodium salt solution 100 mM in H2O](/deepweb/assets/sigmaaldrich/product/structures/352/977/678c0cf4-b036-4984-b198-5f2a4dda679c/640/678c0cf4-b036-4984-b198-5f2a4dda679c.png)
Synonym(s):
dUTP sodium salt
Empirical Formula (Hill Notation):
C9H15N2O14P3 · xNa+
CAS Number:
Molecular Weight:
468.14 (free acid basis)
Products
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing