Skip to Content
Merck
All Photos(1)

Key Documents

APOH316CDE60

Apollo Scientific Ltd

2-Cyclopropyl-4-fluoroaniline

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
151.18
MDL number:

Iupac Name

2-cyclopropyl-4-fluoroaniline

Inchi Code

1S/C9H10FN/c10-7-3-4-9(11)8(5-7)6-1-2-6/h3-6H,1-2,11H2

InChI key

VYZFBDMDLZPLCQ-UHFFFAOYSA-N

storage temp.

Store at -20C

Country of Origin

GB

Shipping Temperature

Shipped with Ice Packs

Physical Form

Solid-Low Melt

Product Link

https://store.apolloscientific.co.uk/product/2-cyclopropyl-4-fluoroaniline

Pictograms

Exclamation mark

Signal Word

Warning

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/2-cyclopropyl-4-fluoroaniline


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service