Merck
Intensified and Continuous Downstream Processing
Gain operational efficiency and flexibility while reducing footprint and costs
Looking to increase your downstream process efficiency?
Transformation of the biomanufacturing workflow is being accelerated by the demand for new therapeutics combined with the need to reduce costs while ensuring operational flexibility and productivity.
Process intensification, closed, and continuous bioprocessing are foundational strategies for achieving these goals. They are widely and successfully deployed in the downstream suite to be more efficient, faster, smaller, less expensive, and sustainable.
To help biopharmaceutical manufacturers intensify, closing, digitally-enhance, or go fully continuous with their processes and gain new levels of operational efficiency and flexibility, there are multiple levers within the DSP suite.
Capture Chromatography
The capture chromatography step is not only a significant contributor to costs, but it can be a bottleneck in the overall DSP (downstream processing) workflow. This downstream purification step can be optimized using continuous multi-column chromatography which considerably reduces the volume and associated cost of Protein A resin required while increasing productivity of the mAb process.
Protein A resins with high binding capacity, superior flow capability, reusability, and a low cost of goods are also essential for process intensification. 
  • Mobius® Multi Column Capture System, a fully automated, single-use 3-column system for continuous closed capture chromatography of your fed-batch or perfusion bioprocess.
  • Eshmuno® A Resin, a rigid, high-capacity Protein A affinity chromatography resin that can be cleaned and sanitized under acid and alkaline conditions while maintaining high dynamic binding capacity at high flow rates.
Related Categories
Operator, fully gowned, standing in front of the Mobius Multi Column Capture System
Chromatography Systems for Bioprocessing
Single-use and multi-use chromatography systems are critical for the separation of your valuable molecules.
4 different sizes of Affinity chromatography with Eshmuno® resin bottles sit on a black counter with a scientist working on a piece of machinery in the background
Affinity Chromatography Resins
Affinity chromatography is often used during the capture phase to isolate the target molecule and reduce process volume.
4 different sizes of Eshmuno CMX, ion exchange chromatography resin bottles
Ion Exchange Chromatography Resins
Ion exchange (IEX) chromatography separates biomolecules based on difference in charge between the target molecule and chromatography resin.
3 sizes of Natrix Q chromatography membrane devices
Membrane Chromatography
Optimize downstream purification using membrane chromatography for capture and polishing. Natrix membranes boost economics and facility efficiency.
Pelicon cassette lab-scale single-pass tangential flow filtration setup
Tangential Flow Filtration Devices
Elevate Filtration Precision: Our tangential flow products ensure purity for mAbs, vaccines, and more, scaling from single-use to multi-use.
Related Resources
Related Webinars
A digital illustration of a laptop displaying a simplistic human figure next to a pink rectangle, all set against a yellow background.
Effective Implementation of Multi Column Chromatography
Improve your process efficiency and reduce costs.
Related Videos
BioContinuum™ Platform: Your enabler of the biomanufacturing facility of the future!