HPLC Analysis of Lipoxidase on TSKgel® Phenyl-5PW
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
TSKgel Phenyl-5PW, 7.5 cm × 7.5 mm I.D., 10 μm particles (807573)
mobile phase
[A] 0.1M phosphate buffer, pH 7.0 + 1.5M ammonium sulfate; [B] 0.1M phosphate buffer, pH 7.0;
gradient
linear, 0% B to 100% B over 60 min
flow rate
0.5 mL/min
detector
UV, 280 nm
Description
Legal Information
TSKgel is a registered trademark of Tosoh Corporation