Merck
  • Surface modification of alumina nanofibres for the selective adsorption of alachlor and imazaquin herbicides.
Surface modification of alumina nanofibres for the selective adsorption of alachlor and imazaquin herbicides.
Journal of colloid and interface science (2011-05-17)
Blain Paul, Wayde N Martens, Ray L Frost
ABSTRACT
The effective removal of pollutants using a thermally and chemically stable substrate that has controllable absorption properties is a goal of water treatment. In this study, the surfaces of thin alumina (γ-Al(2)O(3)) nanofibres were modified by the grafting either of two organosilane agents, 3-chloro-propyl-triethoxysilane (CPTES) and octyl-triethoxysilane (OTES). These modified materials were then trialed as absorbents for the removal of two herbicides, alachlor and imazaquin from water. The formation of organic groups during the functionalisation process established super hydrophobic sites on the surfaces of the nanofibres. This super hydrophobic group is a kind of protruding adsorption site which facilitates the intimate contact with the pollutants. OTES grafted substrate were shown to be more selective for alachlor while imazaquin selectivity is shown by the CPTES grafted substrate. Kinetics studies revealed that imazaquin was rapidly adsorbed on CPTES-modified surfaces. However, the adsorption of alachlor by OTES grafted surface was achieved more slowly.
Product Description
Supelco
Imazaquin, PESTANAL®, analytical standard
Supelco
Alachlor, PESTANAL®, analytical standard