Merck
  • An equine iPSC-based phenotypic screening platform identifies pro- and anti-viral molecules against West Nile virus.
An equine iPSC-based phenotypic screening platform identifies pro- and anti-viral molecules against West Nile virus.
Veterinary research (2024-03-17)
Marielle Cochet, François Piumi, Kamila Gorna, Noémie Berry, Gaëlle Gonzalez, Anne Danckaert, Nathalie Aulner, Odile Blanchet, Stéphan Zientara, Francesc Xavier Donadeu, Hélène Munier-Lehmann, Jennifer Richardson, Alexandra Benchoua, Muriel Coulpier
ABSTRACT
Outbreaks of West Nile virus (WNV) occur periodically, affecting both human and equine populations. There are no vaccines for humans, and those commercialised for horses do not have sufficient coverage. Specific antiviral treatments do not exist. Many drug discovery studies have been conducted, but since rodent or primate cell lines are normally used, results cannot always be transposed to horses. There is thus a need to develop relevant equine cellular models. Here, we used induced pluripotent stem cells to develop a new in vitro model of WNV-infected equine brain cells suitable for microplate assay, and assessed the cytotoxicity and antiviral activity of forty-one chemical compounds. We found that one nucleoside analog, 2'C-methylcytidine, blocked WNV infection in equine brain cells, whereas other compounds were either toxic or ineffective, despite some displaying anti-viral activity in human cell lines. We also revealed an unexpected proviral effect of statins in WNV-infected equine brain cells. Our results thus identify a potential lead for future drug development and underscore the importance of using a tissue- and species-relevant cellular model for assessing the activity of antiviral compounds.
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Sigma-Aldrich
Lovastatin, Lovastatin, CAS 75330-75-5, is an anti-hypercholesterolemic agent that inhibits the activity of 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase.