Merck
  • High miRNA-378 expression has high diagnostic values for pulmonary tuberculosis and predicts adverse outcomes.
High miRNA-378 expression has high diagnostic values for pulmonary tuberculosis and predicts adverse outcomes.
BMC molecular and cell biology (2022-03-21)
Xiaolu Sun, Kai Liu, Yan Zhao, Tianhua Zhang
ABSTRACT
Pulmonary tuberculosis (TB) is a chronic infectious disease. microRNA (miR)-378 is involved in TB diagnosis. This study explored the effects of miR-378 on TB patients. A total of 126 TB patients were selected, including 63 active TB and 63 latent TB, with 62 healthy subjects as controls. Serum miR-378 expression was detected. The diagnostic value of miR-378 in TB was analyzed using the ROC curve. Immune inflammatory factor levels were detected and their correlations with miR-378 expression were analyzed. The drug resistance of active TB patients was recorded after standard treatment. miR-378 expression in drug-resistant TB patients was detected. The effects of miR-378 on adverse outcome incidence were analyzed. miR-378 expression was highly expressed in TB and the expression was higher in the active group than the latent group. Serum miR-378 expression > 1.490 had high sensitivity and specificity in TB diagnosis. miR-378 expression was correlated with TB clinical indexes. IL-4, IL-6, and IL-1β levels were highly expressed, while IFN-γ, TNF-α, and IL-12 levels were lowly expressed in TB patients. Serum miR-378 level in the active group was positively correlated with serum IL-4, IL-6, and IL-1β, and negatively correlated with serum IFN-γ, TNF-α, and IL-12 concentrations. miR-378 expression was downregulated in the TB treated, single (SDR TB) and multi-drug resistance (MDR TB) groups, the miR-378 expression in SDR TB and MDR TB groups was higher than the TB treated group and lower in the SDR TB group than the MDR TB group. High miR-378 expression predicted higher adverse outcome incidence. High miR-378 expression assisted TB diagnosis and predicted adverse outcomes.
Sigma-Aldrich
Human IL-12 p70 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Porcine Tumor Necrosis Factor α ELISA Kit, for serum, plasma and cell culture supernatant