Merck
  • Analytical Detection of Sulfonamides and Organophosphorus Insecticide Residues in Fish in Taiwan.
Analytical Detection of Sulfonamides and Organophosphorus Insecticide Residues in Fish in Taiwan.
Molecules (Basel, Switzerland) (2020-03-29)
Chung-Pei Chang, Po-Hsun Hou, Wei-Cheng Yang, Ching-Fen Wu, Chia-Chia Chang, Ming-Yang Tsai, Hsiao-Pei Tsai, Chien-Teng Lin, Yi-Jing Xue, Jiann-Hsiung Wang, Geng-Ruei Chang
ABSTRACT
Exposure to residues of antibiotics (e.g., sulfonamides) and insecticides (e.g., organophosphorus insecticides) in aquacultured food can adversely affect humans and animals and thus affect public health globally. Here, using a validated method, we examined the levels of residues of 12 sulfonamides as well as 18 organophosphorus insecticides in aquacultured fish in Taiwan. A total of 52 fish samples (i.e., 20 tilapia, 16 milk fish, and 16 perch samples) were obtained from Taiwanese aquafarms from June 2018 to October 2019. We detected 0.02 and 0.03 mg/kg of sulfamethazine (a sulfonamide) in one tilapia and one milk fish, respectively, and 0.02, 0.05, and 0.03 mg/kg of chlorpyrifos (an organophosphorus insecticide) in one tilapia, one milk fish, and one perch, respectively; thus, among the samples, 3.85% and 5.77% contained sulfonamides and organophosphorus insecticide residues, respectively. Furthermore, we assessed human health risk based on the estimated daily intakes (EDIs) of these residues: EDIs of sulfonamide and organophosphorus insecticide residues were <1.0% of the acceptable daily intake recommended by the Joint Food and Agriculture Organization of the United Nations/World Health Organization Expert Committee on Food Additives. The risk of exposure to sulfonamide and organophosphorus insecticide residue by consuming aquacultured fish in Taiwan was thus negligible, signifying no immediate health risk related to the consumption of fish. Our findings can constitute a reference in efforts geared toward ensuring food safety and monitoring veterinary drug and insecticide residue levels in aquacultured organisms. Residue levels in fish must be continually monitored to further determine possible effects of these residues on human health.
Product Description
Supelco
Methacrifos, PESTANAL®, analytical standard
Supelco
Fenamiphos, PESTANAL®, analytical standard