Merck
  • miR-34a, miR-424 and miR-513 inhibit MMSET expression to repress endometrial cancer cell invasion and sphere formation.
miR-34a, miR-424 and miR-513 inhibit MMSET expression to repress endometrial cancer cell invasion and sphere formation.
Oncotarget (2018-05-26)
Peixin Dong, Ying Xiong, Junming Yue, Sharon J B Hanley, Hidemichi Watari
ABSTRACT
Although the oncogene MMSET (also known as NSD2 or WHSC1) has an essential role in malignancies, its impact on human endometrial cancer (EC) metastasis and the molecular mechanism of MMSET regulation are largely unknown. We report that MMSET was markedly upregulated in EC cell lines and EC tissues, and was significantly associated with poor survival in EC. MMSET overexpression greatly promoted EC cell invasion and sphere formation, whereas inhibition of MMSET reduced EC cell invasion and sphere formation. Importantly, Twist1 was required for MMSET-induced EC cell invasion and sphere formation. Moreover, we demonstrate that miR-34a, miR-424 and miR-513 directly modulate MMSET expression to attenuate the invasion and sphere formation capacity of EC cells. miR-34a, miR-424 and miR-513 were down-regulated in EC compared with normal tissue, and reduced expression of miR-34a, miR-424 and miR-513 was clinically associated with a poorer prognosis in EC patients. Furthermore, specific inhibition of MMET with BIX-01294 led to decreased EC cell invasion and impaired sphere formation. These findings suggest a pro-metastatic role for MMSET in EC and reveal that the repression of miR-34a, miR-424 and miR-513 contributes to the overexpression of MMSET during EC metastasis.
Sigma-Aldrich
Anti-NSD2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available. Contact Technical Service