
Sumatriptan for system suitability

Synonym(s):
Sumatriptan succinate, 3-[2-(Dimethylamino)ethyl]-N-methyl-1H-indole-5-methanesulfonamide succinate, GR-43175
Empirical Formula (Hill Notation):
C14H21N3O2S · C4H6O4
CAS Number:
Molecular Weight:
413.49
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing