
Pinoxaden Metabolite SYN505164

Synonym(s):
8-[2,6-Diethyl-4-(hydroxymethyl)phenyl]tetrahydropyrazolo[1,2-d][1,4,5]oxadiazepine-7-9-dione
Empirical Formula (Hill Notation):
C18H24N2O4
CAS Number:
Molecular Weight:
332.39
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing