
Penicillin G benzathine tetrahydrate

Empirical Formula (Hill Notation):
C32H36N4O8S2 · C16H20N2 · 4H2O
CAS Number:
Molecular Weight:
981.18
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing