
Myricetin 3-O-β-D-Galactopyranoside

Empirical Formula (Hill Notation):
C21H20O13
CAS Number:
Molecular Weight:
480.38
Products
Product Category
Brand
Formula Weight
Physical Form
Research Area
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing