
Moexipril Related Compound A

Synonym(s):
Moexiprilat, (3S)-2-{(2S)-N-[(1S)-1-Carboxy-3-phenylpropyl]alanyl}-1,2,3,4-tetrahydro-6,7-dimethoxy-3-isoquinolinecarboxylic acid
Empirical Formula (Hill Notation):
C25H30N2O7
CAS Number:
Molecular Weight:
470.51
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2