Skip to Content
Merck
L-Pyroglutamic acid BioXtra

L-Pyroglutamic acid

L-Pyroglutamic acid BioXtra
Synonym(s):
(S)-(−)-2-Pyrrolidone-5-carboxylic acid, (S)-5-Oxo-2-pyrrolidinecarboxylic acid
Empirical Formula (Hill Notation):
C5H7NO3
CAS Number:
Molecular Weight:
129.11
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Special Grade
Technique