
Herbacetin

Synonym(s):
3,4′,5,7,8-Pentahydroxyflavone, 3,5,7,8-Tetrahydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 4′,5,7,8-Tetrahydroxyflavonol, 8-Hydroxykaempferol, Isoarticulatidin
Empirical Formula (Hill Notation):
C15H10O7
CAS Number:
Molecular Weight:
302.24
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing